Learn More
Alkali Scientific Tryptic Soy Broth (TSB) 90mL Bottle, 1/Pk
SDP
Catalog No. 5000662464
Encompass_Preferred
Tryptic Soy Broth (TSB) is a general purpose liquid culture medium widely used for the cultivation of a broad range of microorganisms, including bacteria and fungi. This 90mL sterile bottle format is specifically designed for dilution preparation and sample processing in microbiological workflows. The nutrient rich formulation contains enzymatic digests of casein and soybean meal, providing essential amino acids, vitamins, and growth factors that support rapid microbial growth. It is commonly used for preparing sample dilutions, pre enrichment of microorganisms, and routine culture work. The ready to use sterile format reduces preparation time and contamination risk, making it suitable for laboratory testing, quality control, and environmental monitoring applications.
Catalog No. 50-006-62464 Supplier Alkali Scientific Supplier No. TSBB05
Tryptic Soy Broth (TSB) is a general purpose liquid culture medium widely used for the cultivation of a broad range of microorganisms, including bacteria and fungi. This 90mL sterile bottle format is specifically designed for dilution preparation and sample processing in microbiological workflows. The nutrient rich formulation contains enzymatic digests of casein and soybean meal, providing essential amino acids, vitamins, and growth factors that support rapid microbial growth. It is commonly used for preparing sample dilutions, pre enrichment of microorganisms, and routine culture work. The ready to use sterile format reduces preparation time and contamination risk, making it suitable for laboratory testing, quality control, and environmental monitoring applications.