Learn More
Alkali Scientific Tryptic Soy Broth (TSB) 9mL Tube, 10/Pk
SDP
Catalog No. 5000662470
Encompass_Preferred
Tryptic Soy Broth (TSB) is a versatile liquid culture medium used for the growth and maintenance of a wide variety of microorganisms. This 9mL sterile tube format is specifically designed for serial dilution procedures in microbiology laboratories. By adding a measured volume of sample to the tube, technicians can easily prepare stepwise dilutions for microbial enumeration and testing. The nutrient rich composition supports consistent microbial recovery while maintaining sample integrity during processing. These tubes are commonly used in microbiological analysis, quality control testing, and research workflows where accurate dilution preparation and reliable growth conditions are required.
Catalog No. 50-006-62470 Supplier Alkali Scientific Supplier No. TSBT04
Tryptic Soy Broth (TSB) is a versatile liquid culture medium used for the growth and maintenance of a wide variety of microorganisms. This 9mL sterile tube format is specifically designed for serial dilution procedures in microbiology laboratories. By adding a measured volume of sample to the tube, technicians can easily prepare stepwise dilutions for microbial enumeration and testing. The nutrient rich composition supports consistent microbial recovery while maintaining sample integrity during processing. These tubes are commonly used in microbiological analysis, quality control testing, and research workflows where accurate dilution preparation and reliable growth conditions are required.