Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Hardy Diagnostics™ Tryptic Soy Broth (TSB), Double Strength
SDP

Catalog No. p-200005338 Shop All Hardy Diagnostics Products
Change view
Click to view available options
Fill Volume:
25 mL
50 mL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Fill Volume
23-001-824 50 mL
23-001-784 25 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Catalog No. 23-001-824 Supplier Hardy Diagnostics™ Supplier No. U205
Only null left
Add to Cart
Add to Cart
This item is not returnable. View return policy

Tryptic Soy Broth (TSB), Double Strength is recommended for use as a general purpose medium for the isolation and cultivation of a wide variety of bacteria and fungi

  • Tryptic Soy Broth is widely used for the isolation of bacteria from clinical specimens, supporting the growth of a majority of pathogenic bacteria
  • This medium contains digests of soybean meal and casein, which provide amino acids and other nitrogenous substances, making it a highly nutritious medium for a variety of organisms, including most pathogenic bacteria
  • Dipotassium phosphate is included in the formulation as a buffer to maintain the pH

Specifications

Appearance Clear
Color Medium Amber
Form Liquid
Product Type Broth Tube
Fill Volume 50 mL
Incubation Conditions 20°C to 25°C, 30°C to 35°C
Incubation Time 1 to 5 days
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.