Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Hardy Diagnostics Tryptic Soy Broth (TSB), USP, 10 mL Fill, Vial with Needle Port Septum
SDP

Catalog No. 22026377 Shop All Hardy Diagnostics Products
Change view
Click to view available options
Target Organism Class:
Bacteria, Yeast, Molds
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Target Organism Class
22-026-377 Bacteria, Yeast, Molds
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 22-026-377 Supplier Hardy Diagnostics Supplier No. U8210
Add to Cart
Add to Cart

It is recommended for use as a general purpose medium for the isolation and cultivation of a wide variety of bacteria and fungi.

  • The U.S. Pharmacopeia National Formulary (USP) describes its use for sterility testing
  • Sterility media and rinse solutions are manufactured in an ISO 13485 environmentally controlled production center
  • Each lot is certified for pH, sterility and growth promotion using ATCC™ strains specified by the USP
  • Our approach to manufacturing ensures all sterility media and rinse fluids provide the highest level of clarity for improved reading accuracy, thus significantly reducing the risk of incorrect interpretation and false results

Specifications

Form Liquid
Manufacturing Origin US with Globally Sourced Materials
Product Type Broth, Liquid Medium
Sterility Non-sterile
Target Organism Class Bacteria, Yeast, Molds
Test Type Microbial Detection, Media Testing, Growth Promotion Testing, Media Testing
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.