Learn More
Hardy Diagnostics Tryptic Soy Broth (TSB), USP, 100 mL Fill, Wide Mouth Polycarbonate Jar
Shop All Hardy Diagnostics Products

Description
- The U.S. Pharmacopeia National Formulary (USP) describes its use for sterility testing
- Bottle features:
- Heavy-duty: Bottles are tough and sturdy to reduce breakage
- Optically clear: Bottles are made of clarified polycarbonate for clear viewing
Specifications
Specifications
| Form | Liquid |
| Manufacturing Origin | US with Globally Sourced Materials |
| Product Type | Broth, Liquid Medium |
| Sterility | Non-sterile |
| Target Organism Class | Bacteria, Molds, Yeast |
| Test Type | Microbial Detection, Media Testing, Growth Promotion Testing |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.