Learn More
Alkali Scientific Tryptic Soy Broth (TSB) w/0.1% Sodium Thiosulfate 200mL Bottle, 12/CS
SDP
Catalog No. 5000662467
Encompass_Preferred
Tryptic Soy Broth (TSB) with 0.1% Sodium Thiosulfate 200mL Bottle, 12/CS. This specialized broth formulation includes sodium thiosulfate, ideal for microbial neutralization and testing. It offers chemical-resistant properties and enhances the neutralizing performance for various microorganisms. The 200mL bottle size is perfect for high-throughput lab applications. Comes in a 12-pack for larger scale experiments and microbial testing.
Catalog No. 50-006-62467 Supplier Alkali Scientific Supplier No. TSBB12
Tryptic Soy Broth (TSB) with 0.1% Sodium Thiosulfate 200mL Bottle, 12/CS. This specialized broth formulation includes sodium thiosulfate, ideal for microbial neutralization and testing. It offers chemical-resistant properties and enhances the neutralizing performance for various microorganisms. The 200mL bottle size is perfect for high-throughput lab applications. Comes in a 12-pack for larger scale experiments and microbial testing.