Learn More
Alkali Scientific Tryptic Soy Broth (TSB) w/0.5% Lecithin & 4% Tween 80 990mL Bottle, 1/PK
SDP
Catalog No. 5000663271
Encompass_Preferred
Tryptic Soy Broth (TSB) with 0.5% Lecithin & 4% Tween 80, 990mL Bottle, 1/PK, is used as a neutralizing agent in microbial growth studies. It is designed for use in laboratory applications, providing optimal conditions for microorganism survival and testing. The lecithin and Tween 80 provide effective neutralization of preservatives and disinfectants, ensuring that results are accurate and reproducible for testing in various microbiological fields.
Catalog No. 50-006-63271 Supplier Alkali Scientific Supplier No. TSBB07
Tryptic Soy Broth (TSB) with 0.5% Lecithin & 4% Tween 80, 990mL Bottle, 1/PK, is used as a neutralizing agent in microbial growth studies. It is designed for use in laboratory applications, providing optimal conditions for microorganism survival and testing. The lecithin and Tween 80 provide effective neutralization of preservatives and disinfectants, ensuring that results are accurate and reproducible for testing in various microbiological fields.