Learn More
Alkali Scientific Tryptic Soy Broth (TSB) w/.5 gm Lecithin & 40mL Tween 20/L 990mL Bottle, 1/PK
SDP
Catalog No. 5000662854
Encompass_Preferred
Tryptic Soy Broth with Lecithin and Tween 20 is a specialized neutralizing medium used in environmental monitoring and microbiological testing. The formulation includes lecithin and Tween 20, which act as neutralizing agents to inactivate disinfectants and antimicrobial residues commonly found on surfaces or in samples. This allows microorganisms that may have been exposed to cleaning agents to recover and grow during testing. The 990mL bottle provides a large working volume suitable for bulk sample processing or preparation of test solutions. This medium is widely used in pharmaceutical, food, and industrial laboratories where accurate detection of microbial contamination is required despite the presence of inhibitory substances.
Catalog No. 50-006-62854 Supplier Alkali Scientific Supplier No. TSBB06
Tryptic Soy Broth with Lecithin and Tween 20 is a specialized neutralizing medium used in environmental monitoring and microbiological testing. The formulation includes lecithin and Tween 20, which act as neutralizing agents to inactivate disinfectants and antimicrobial residues commonly found on surfaces or in samples. This allows microorganisms that may have been exposed to cleaning agents to recover and grow during testing. The 990mL bottle provides a large working volume suitable for bulk sample processing or preparation of test solutions. This medium is widely used in pharmaceutical, food, and industrial laboratories where accurate detection of microbial contamination is required despite the presence of inhibitory substances.