Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Innovating Science™ Tryptic Soy Broth Tubes
SCI_ED

Catalog No. S802483 Shop All Aldon Products
Change view
Click to view available options
No. per Pack:
12 Pk.
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. No. per Pack
S802483 12 Pk.
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Catalog No. S802483 Supplier Innovating Science™ Supplier No. IS5361
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Tryptic Soy Broth Tubes are manufactured sterile microbiological media for the lab and education market. The classic black rubber lined phenolic caps ensure a sterile seal. Great quality and competitive pricing will help you outsource or upgrade your current product line.

  • Phenolic caps ensure sterile seal
  • Great quality
  • Competitive pricing

Specifications

Detects Bacterial Growth
Form Liquid
Format 6 mL Tubes
Grade 9 to 12
Manufacturing Origin United States
Media Type Tryptic Soy Broth
Preparation Method Autoclave
Product Type Broth Tube
Sterility Sterile
Target Organism Class Fastidious and Non-fastidious Organisms
Test Type Microbial Detection
For Use With (Application) Growing Bacteria
No. per Pack 12 Pk.
Packaging Type Box
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.