Learn More
Alkali Scientific Tryptic Soy Broth w/2% Glycerol 1mL, 2mL Vial, 500/Bag
SDP
Catalog No. 5000662816
Encompass_Preferred
Tryptic Soy Broth (TSB) with 2% glycerol is a nutrient rich medium designed for the cultivation and cryopreservation of a wide range of bacterial strains. The addition of glycerol functions as a cryoprotectant, helping maintain cell viability during freezing and long term storage. Supplied with a 1mL fill in sterile 2mL vials, this ready to use format provides consistent aliquots suitable for microbial culture storage, strain preservation, and biobanking applications. The medium supports growth of aerobic and facultative anaerobic organisms and is widely used in research, clinical, and industrial microbiology laboratories. It is ideal for microbial recovery, culture maintenance, and long term preservation of laboratory strains.
Catalog No. 50-006-62816 Supplier Alkali Scientific Supplier No. TSAT05
Tryptic Soy Broth (TSB) with 2% glycerol is a nutrient rich medium designed for the cultivation and cryopreservation of a wide range of bacterial strains. The addition of glycerol functions as a cryoprotectant, helping maintain cell viability during freezing and long term storage. Supplied with a 1mL fill in sterile 2mL vials, this ready to use format provides consistent aliquots suitable for microbial culture storage, strain preservation, and biobanking applications. The medium supports growth of aerobic and facultative anaerobic organisms and is widely used in research, clinical, and industrial microbiology laboratories. It is ideal for microbial recovery, culture maintenance, and long term preservation of laboratory strains.