Learn More
Teknova Tryptic Soy Broth with Animal-Free Soytone, 1000mL, Sterile
SDP
Catalog No. 502388096
Encompass
Tryptic soy media (TSM) is a general purpose medium used for the isolation and enumeration of enteric bacteria in food and water samples. This medium is also used in sterility testing, as it provides optimum growth conditions for a variety of bacteria.
Catalog No. 50-238-8096 Supplier Teknova Supplier No. T8510
Tryptic soy media (TSM) is a general purpose medium used for the isolation and enumeration of enteric bacteria in food and water samples. This medium is also used in sterility testing, as it provides optimum growth conditions for a variety of bacteria.