Learn More
MilliporeSigma Tryptic Soy Agar - ICR 30 ml (03075e) 120 pack
Catalog No. NC9077395
Encompass
Application
Tryptic Soy Agar (TSA, Soybean Casein Digest Agar) is a complex medium for cultivation and isolation of a wide range of bacteria, yeasts and molds. The media finds its application in environmental monitoring by determining the total aerobic microbial count in air through active or passive air monitoring as well as fingerprints of personnel in isolators and clean rooms
Analysis Note
Tryptic Soy Agar - ICR settle plate, is a non-lockable plate and should be used for aerobic incubation only
This item is not returnable. View return policy
Catalog No. NC9077395 Supplier MilliporeSigma Supplier No. 1460010120
Application
Tryptic Soy Agar (TSA, Soybean Casein Digest Agar) is a complex medium for cultivation and isolation of a wide range of bacteria, yeasts and molds. The media finds its application in environmental monitoring by determining the total aerobic microbial count in air through active or passive air monitoring as well as fingerprints of personnel in isolators and clean rooms
Analysis Note
Tryptic Soy Agar - ICR settle plate, is a non-lockable plate and should be used for aerobic incubation only