Learn More
Teknova Trypticase Soy Agar Plates, 100mm Plates, 20 Plates/Sleeve, Sterile.
SDP
Catalog No. 502388000
Encompass
Tryptic soy media (TSM) is a general purpose medium used for the isolation and enumeration of enteric bacteria in food and water samples. This medium is also used in sterility testing, as it provides optimum growth conditions for a variety of bacteria.
Catalog No. 50-238-8000 Supplier Teknova Supplier No. T1144
Tryptic soy media (TSM) is a general purpose medium used for the isolation and enumeration of enteric bacteria in food and water samples. This medium is also used in sterility testing, as it provides optimum growth conditions for a variety of bacteria.