Learn More
Teknova Trypticase Soy Broth (TSB), 450 ml 8 pack.
SDP
Catalog No. 502387668
Encompass
Tryptic soy media (TSM) is a general purpose medium used for the isolation and enumeration of enteric bacteria in food and water samples. This medium is also used in sterility testing, as it provides optimum growth conditions for a variety of bacteria.
Catalog No. 50-238-7668 Supplier Teknova Supplier No. T58218PK
Tryptic soy media (TSM) is a general purpose medium used for the isolation and enumeration of enteric bacteria in food and water samples. This medium is also used in sterility testing, as it provides optimum growth conditions for a variety of bacteria.