Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Research Products International Corp Trypto Soy Agar, 5 KG
SDP

Catalog No. 502230459
Encompass_Preferred

Trypto Soy Agar is a general growth medium prepared with or without blood or enrichments. It is used for the isolation of different strains of fastidious microorganisms.

A blood agar medium may be prepared by adding 5 to 10% sterile defibrinated blood. TSA with 5% sheep blood is useful for the isolation and determination of hemolytic reactions of staphylococcus sp., steptococcus sp. and other pathogenic microorganisms found in the upper lower respiratory tract, blood, cerebrospinal fluid, minor lesions and other clinical specimens.

Formula per Liter:

Casein Digest Peptone: 15.0g
Agar: 15.0g
Papaic Digest of Soybean Meal: 5.0g
Sodium Chloride: 5.0g
Final pH: 7.3 ± 0.2 at 25.0°C

Catalog No. 50-223-0459 Supplier Research Products International Corp Supplier No. T488005000.0
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Trypto Soy Agar is a general growth medium prepared with or without blood or enrichments. It is used for the isolation of different strains of fastidious microorganisms.

A blood agar medium may be prepared by adding 5 to 10% sterile defibrinated blood. TSA with 5% sheep blood is useful for the isolation and determination of hemolytic reactions of staphylococcus sp., steptococcus sp. and other pathogenic microorganisms found in the upper lower respiratory tract, blood, cerebrospinal fluid, minor lesions and other clinical specimens.

Formula per Liter:

Casein Digest Peptone: 15.0g
Agar: 15.0g
Papaic Digest of Soybean Meal: 5.0g
Sodium Chloride: 5.0g
Final pH: 7.3 ± 0.2 at 25.0°C

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.