Learn More
Research Products International Corp Trypto Soy Broth, 1 KG

Supplier: Research Products International Corp T485001000.0
Dissolve 30 grams in 1 liter of distilled water. Sterilize by autoclaving for 15 minutes.
Formula Per Liter:
Casein Digest Peptone: 17.0 g
Papaic Digest of Soybean Meal: 3.0 g
Dipotassium Phosphate: 2.5 g
Dextrose: 2.5 g
Sodium Chloride: 5.0 g
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.