Learn More
Research Products International Corp Tryptone, 1 Gram Capsules, 500 Grams
SDP
Catalog No. 50550444
Encompass_Preferred
Each capsule contains 1g Tryptone. Convenient and easy to use - simply add required number of premeasured capsules according to formulation being prepared without weighing or measuring.
Catalog No. 50-550-444 Supplier Research Products International Corp Supplier No. T60061500.0
Each capsule contains 1g Tryptone. Convenient and easy to use - simply add required number of premeasured capsules according to formulation being prepared without weighing or measuring.