Learn More
Bioworld Tryptone - Casein Peptone Type T, 500 g
Catalog No. 50254539
Encompass_Preferred
TryptoneEnzymatic digest of casein used in preparing microbiological culture mediaA rich source of amino acids and peptidesTechnical Specifications:Pancreatic Digest of Casein Total Nitrogen (TN): 12.7%Amino Nitrogen (AN): 3.7%AN/TN Ratio: 0.26%Sodium Chloride: 0.4%Tryptophan: 1.0%pH (1% solution): 7.3Soluble in water
Catalog No. 50-254-539 Supplier Bioworld Supplier No. 306270252
TryptoneEnzymatic digest of casein used in preparing microbiological culture mediaA rich source of amino acids and peptidesTechnical Specifications:Pancreatic Digest of Casein Total Nitrogen (TN): 12.7%Amino Nitrogen (AN): 3.7%AN/TN Ratio: 0.26%Sodium Chloride: 0.4%Tryptophan: 1.0%pH (1% solution): 7.3Soluble in water