Learn More
IBI SCIENTIFIC 3P Tryptone, a Pancreatic Digest of Casein, 500 g
Catalog No. NC0753542
Encompass_Preferred
Tryptone - 500gm Tryptone is a pancreatic digest of casein and used as a nitrogen source in culture media. It is capable of supporting growth of fastidious and nonfastidious micro- organisms.
Catalog No. NC0753542 Supplier IBI SCIENTIFIC 3P Supplier No. IB49180
Tryptone - 500gm Tryptone is a pancreatic digest of casein and used as a nitrogen source in culture media. It is capable of supporting growth of fastidious and nonfastidious micro- organisms.