Learn More
Medchemexpress LLC Tryptone | 73049-73-7 | MFCD00131829 | 25 G
SDP
Catalog No. 5000283368
Encompass_Preferred
Tryptone is a peptide-rich enzymatic hydrolysate used as a carbon and nitrogen source in bacterial culture media. It supplies oligopeptides that influence bacterial surface proteins and biofilm-related gene expression, and is commonly used to support bacterial growth and biofilm or adhesion studies.
  • Peptide-rich carbon and nitrogen source for bacterial culture.
  • Supports biofilm and adhesion studies by providing oligopeptides that affect surface protein expression.
  • Solid powder, yellow to brown in color; suitable for room temperature storage.
  • Available in 25 g, 50 g, and larger pack sizes.
  • CAS number 73049-73-7.
Catalog No. 50-002-83368 Supplier Medchemexpress LLC Supplier No. HYW13389825G
Tryptone is a peptide-rich enzymatic hydrolysate used as a carbon and nitrogen source in bacterial culture media. It supplies oligopeptides that influence bacterial surface proteins and biofilm-related gene expression, and is commonly used to support bacterial growth and biofilm or adhesion studies.
  • Peptide-rich carbon and nitrogen source for bacterial culture.
  • Supports biofilm and adhesion studies by providing oligopeptides that affect surface protein expression.
  • Solid powder, yellow to brown in color; suitable for room temperature storage.
  • Available in 25 g, 50 g, and larger pack sizes.
  • CAS number 73049-73-7.