Learn More
LAMDA BIOTECH INC Tryptone
Catalog No. 502045830
Encompass_Preferred
High quality peptone, molecular biology certified (MBC). This product is a pancreatic digest of casein. It is widely used for general bacteriological research work with a variety of microorganisms, such as for making various microbiological media and as an additive for other culture media.
Catalog No. 50-204-5830 Supplier LAMDA BIOTECH INC Supplier No. C109
High quality peptone, molecular biology certified (MBC). This product is a pancreatic digest of casein. It is widely used for general bacteriological research work with a variety of microorganisms, such as for making various microbiological media and as an additive for other culture media.