Learn More
Bioworld Tryptone-Azolectin-Tween (TAT) Broth Base
Catalog No. 50254373
Encompass
Tryptone-Azolectin-Tween™ (TAT) Broth BaseUsed for cultivating microorganisms from highly viscous or gelatinous materialspH: 7.2Working Concentration: 25 g/LDissolve in 960 mL purified water with 40 mL polysorbate 20Dehydrated Appearance: beige, homogeneous powderPrepared Appearance: light amber brothSoluble in warm water/polysorbate 20 mixtureContains:Pancreatic Digest of Casein Lecithin
Catalog No. 50-254-373 Supplier Bioworld Supplier No. 306200801
Tryptone-Azolectin-Tween™ (TAT) Broth BaseUsed for cultivating microorganisms from highly viscous or gelatinous materialspH: 7.2Working Concentration: 25 g/LDissolve in 960 mL purified water with 40 mL polysorbate 20Dehydrated Appearance: beige, homogeneous powderPrepared Appearance: light amber brothSoluble in warm water/polysorbate 20 mixtureContains:Pancreatic Digest of Casein Lecithin