Learn More
Crescent Chemical Co Inc TRYPTONE FROM CASEIN PANCREATI
SDP
Supplier:  Crescent Chemical Co Inc SE48647.01
Encompass_Preferred
Tryptone from casein pancreatic Prepared by pancreatic digest of milk protein casein. Very rich source of amino nitrogen. Used in the production of various general media such as Tryptone Water etc. and as well in the production of sterility testing media and various
Catalog No. 50-249-171