Learn More
Bioworld Tryptone Glucose Extract (TGE) Agar, 500 g
Catalog No. 50254387
Encompass_Preferred
Tryptone Glucose Extract (TGE) AgarFor the cultivation and enumeration of bacteria from milk and dairy products Meets APHA specifications for obtaining the microbial content of food, dairy products, water and wastewaterpH: 7.0Working Concentration: 24 g/LDehydrated Appearance: light to medium tan, homogeneous powderPrepared Appearance: light amber agarSoluble in boiling waterContains:Tryptone Beef Extract Dextrose Agar
Catalog No. 50-254-387 Supplier Bioworld Supplier No. 306200881
Tryptone Glucose Extract (TGE) AgarFor the cultivation and enumeration of bacteria from milk and dairy products Meets APHA specifications for obtaining the microbial content of food, dairy products, water and wastewaterpH: 7.0Working Concentration: 24 g/LDehydrated Appearance: light to medium tan, homogeneous powderPrepared Appearance: light amber agarSoluble in boiling waterContains:Tryptone Beef Extract Dextrose Agar