Learn More
NEOGEN Tryptone No. 1, 10 kg
SDP
Catalog No. 502015749
Encompass_Preferred
Tryptone No. 1 is an enzymatic digest of casein and meat for use in preparing microbiological culture media in a laboratory setting. Tryptone No. 1 is not intended for use in the diagnosis of disease or other conditions in humans. Tryptone No. 1 is an enzymatic digest of casein and meat used as a nitrogen source in culture media. Casein is the main protein of milk, and a rich source of amino-acid nitrogen. Tryptone No. 1 is rich in tryptophane, making it valuable for use in detecting indole production. The absence of detectable levels of carbohydrates in Tryptone makes it a suitable peptone in differentiating bacteria on the basis of their ability to ferment various carbohydrates. Several media containing Tryptone are specified in standard methods for multiple applications.For more information visit https://www.neogen.com/solutions/Microbiology/tryptone-number-1
Catalog No. 50-201-5749 Supplier NEOGEN Supplier No. 700003524
Tryptone No. 1 is an enzymatic digest of casein and meat for use in preparing microbiological culture media in a laboratory setting. Tryptone No. 1 is not intended for use in the diagnosis of disease or other conditions in humans. Tryptone No. 1 is an enzymatic digest of casein and meat used as a nitrogen source in culture media. Casein is the main protein of milk, and a rich source of amino-acid nitrogen. Tryptone No. 1 is rich in tryptophane, making it valuable for use in detecting indole production. The absence of detectable levels of carbohydrates in Tryptone makes it a suitable peptone in differentiating bacteria on the basis of their ability to ferment various carbohydrates. Several media containing Tryptone are specified in standard methods for multiple applications.For more information visit https://www.neogen.com/solutions/Microbiology/tryptone-number-1