Learn More
MOLTOX Molecular Toxicology Inc Tryptone Salt Broth, 9 mL/tube, 10 tubes/box, Store at Room Temperature
SDP
Catalog No. 50948841
Encompass_Preferred
This Tryptone Salt Broth is a 0.1% tryptone with NaCl for use as a diluent in stock suspensions. Each lot of media from MOLTOX is tested for sterility, where applicable, and its ability to support the growth of the appropriate microorganism. All media are accompanied by GLP-compliant formulation and Quality Control certificate.
Catalog No. 50-948-841 Supplier MOLTOX Molecular Toxicology Inc Supplier No. 5140S100
This Tryptone Salt Broth is a 0.1% tryptone with NaCl for use as a diluent in stock suspensions. Each lot of media from MOLTOX is tested for sterility, where applicable, and its ability to support the growth of the appropriate microorganism. All media are accompanied by GLP-compliant formulation and Quality Control certificate.