Learn More
NEOGEN Tryptone Soy Agar, 500 g
SDP
Catalog No. 502015210
Encompass_Preferred
Tryptone Soy Agar (TSA) is a general-purpose agar which will support the growth of a wide range of micro-organisms, and is not intended for use in the diagnosis of disease or other conditions in humans.The medium can be used for phage typing, colicine typing and for testing the X and V factor requirements of Haemophilus spp.Formula:Tryptone - 15.0 g/LSoy Peptone - 0.5 g/LSodium Chloride - 5.0 g/LAgar - 12.0 g/LFor more information visit https://www.neogen.com/solutions/Microbiology/tryptone-soy-agar
Catalog No. 50-201-5210 Supplier NEOGEN Supplier No. 700004389
Tryptone Soy Agar (TSA) is a general-purpose agar which will support the growth of a wide range of micro-organisms, and is not intended for use in the diagnosis of disease or other conditions in humans.The medium can be used for phage typing, colicine typing and for testing the X and V factor requirements of Haemophilus spp.Formula:Tryptone - 15.0 g/LSoy Peptone - 0.5 g/LSodium Chloride - 5.0 g/LAgar - 12.0 g/LFor more information visit https://www.neogen.com/solutions/Microbiology/tryptone-soy-agar