Learn More
Medchemexpress LLC Tryptophan Hydroxyla 50ug
SDP
Catalog No. 5000180060
Encompass_Preferred
The tryptophan hydroxylase 1 (TPH-1) protein is key to serotonin biosynthesis catalyzing the rate-limiting step the oxidation of L-tryptophan to 5-hydroxy-L-tryptophan This enzyme activity is critical for the synthesis of serotonin a neurotransmitter that affects mood sleep and appetite Tryptophan Hydroxylase 1/TPH-1 Protein Human (His) is the recombinant human-derived Tryptophan Hydroxylase 1/TPH-1 protein expressed by E coli with N-His labeled tag
Catalog No. 50-001-80060 Supplier Medchemexpress LLC Supplier No. HYP7356650UG
The tryptophan hydroxylase 1 (TPH-1) protein is key to serotonin biosynthesis catalyzing the rate-limiting step the oxidation of L-tryptophan to 5-hydroxy-L-tryptophan This enzyme activity is critical for the synthesis of serotonin a neurotransmitter that affects mood sleep and appetite Tryptophan Hydroxylase 1/TPH-1 Protein Human (His) is the recombinant human-derived Tryptophan Hydroxylase 1/TPH-1 protein expressed by E coli with N-His labeled tag