Learn More
CPC Scientific H-Gly-Gly-Trp-Ser-His-Trp-NH2 (trifluoroacetate salt) 1MG
SDP
Catalog No. 502110560
Encompass
This fibronectin-binding peptide, derived from the second type I repeat of thrombospondin, is an inhibitor of the fibronectin-mediated interactions of cells with collagen in the extracellular matrix. It is completely conserved in the second type I repeats of mouse and human thrombospondin-1 and thrombospondin-2 from mouse and chickenSEQUENCE: H-Gly-Gly-Trp-Ser-His-Trp-NH2 (trifluoroacetate salt)ONE-LETTER SEQUENCE: GGWSHW-NH2MOLECULAR FORMULA: C35H41N11O7MOLECULAR WEIGHT: 727.79STORAGE CONDITIONS: -20 5 CCAS REGISTRY NUMBER: SYNONYMS: RESEARCH AREA: HematologyREFERENCES:Sipes, J. J. Cell Biol. 121, 469 (1993)Shultz-Cherry, S. et al. J. Biol. Chem. 270, 7304 (1995)SKU(s): THBO-003A
Catalog No. 50-211-0560 Supplier CPC Scientific Supplier No. THBO003A
Add to Cart
Add to Cart
This fibronectin-binding peptide, derived from the second type I repeat of thrombospondin, is an inhibitor of the fibronectin-mediated interactions of cells with collagen in the extracellular matrix. It is completely conserved in the second type I repeats of mouse and human thrombospondin-1 and thrombospondin-2 from mouse and chickenSEQUENCE: H-Gly-Gly-Trp-Ser-His-Trp-NH2 (trifluoroacetate salt)ONE-LETTER SEQUENCE: GGWSHW-NH2MOLECULAR FORMULA: C35H41N11O7MOLECULAR WEIGHT: 727.79STORAGE CONDITIONS: -20 5 CCAS REGISTRY NUMBER: SYNONYMS: RESEARCH AREA: HematologyREFERENCES:Sipes, J. J. Cell Biol. 121, 469 (1993)Shultz-Cherry, S. et al. J. Biol. Chem. 270, 7304 (1995)SKU(s): THBO-003A