Learn More
U.S. Pharmacopeia Tryptophan Related Compound B, 1218-34-4, MFCD00065976, 25mg
Supplier:  U.S. Pharmacopeia 1700523
Encompass
Molecular Formula C13 H14 N2 O3, Molecular weight 246.29, Melting point 364.28 - 373.1 °F (184.6 - 189.5 °C), Synonyms: Acetyltryptophan, N-Acetyl-L-tryptophan
Catalog No. NC0006826