Learn More
Medchemexpress LLC Tryptophan Synthase 1mg
SDP
Catalog No. 5000196772
Encompass_Preferred
Recombinant E. coli tryptophan synthase (TrpA/TrpB) supplied as a filtered PBS solution with an N-terminal 6xHis tag. The dimeric enzyme catalyzes the conversion of indole glycerol phosphate to indole and glyceraldehyde-3-phosphate and is provided for biochemical and enzymatic assays; it is shipped on dry ice and intended for research use with recommended frozen storage.
Catalog No. 50-001-96772 Supplier Medchemexpress LLC Supplier No. HYP710981MG
Recombinant E. coli tryptophan synthase (TrpA/TrpB) supplied as a filtered PBS solution with an N-terminal 6xHis tag. The dimeric enzyme catalyzes the conversion of indole glycerol phosphate to indole and glyceraldehyde-3-phosphate and is provided for biochemical and enzymatic assays; it is shipped on dry ice and intended for research use with recommended frozen storage.