Learn More
Bioworld Tryptose Agar, 500 g
Catalog No. 50254389
Encompass_Preferred
Tryptose AgarFor cultivating a wide variety of fastidious microorganisms, particularly for isolating BrucellapH: 7.2Working Concentration: 41 g/LDehydrated Appearance: light beige, homogeneous powderPrepared Appearance: light amber agarSoluble in boiling waterContains:Tryptose Sodium Chloride Dextrose Agar
Catalog No. 50-254-389 Supplier Bioworld Supplier No. 306200911
Tryptose AgarFor cultivating a wide variety of fastidious microorganisms, particularly for isolating BrucellapH: 7.2Working Concentration: 41 g/LDehydrated Appearance: light beige, homogeneous powderPrepared Appearance: light amber agarSoluble in boiling waterContains:Tryptose Sodium Chloride Dextrose Agar