Learn More
NEOGEN Tryptose No. 2, 500 g
SDP
Catalog No. 502015876
Encompass_Preferred
Tryptose No. 2 is a special blend of peptones developed for use in the production of Foot and Mouth vaccine, and is not intended for use in the diagnosis of disease or other conditions in humans.A special blend of peptones originally developed for use in the production of Foot and Mouth vaccine. Other peptone blends are also available within the catalogue, some of which have been evaluated to demonstrate low cell toxicity and the ability to enhance the growth of cell lines (eg NCM0264).For more information visit https://www.neogen.com/solutions/Microbiology/tryptose-number-2
Catalog No. 50-201-5876 Supplier NEOGEN Supplier No. 700004744
Tryptose No. 2 is a special blend of peptones developed for use in the production of Foot and Mouth vaccine, and is not intended for use in the diagnosis of disease or other conditions in humans.A special blend of peptones originally developed for use in the production of Foot and Mouth vaccine. Other peptone blends are also available within the catalogue, some of which have been evaluated to demonstrate low cell toxicity and the ability to enhance the growth of cell lines (eg NCM0264).For more information visit https://www.neogen.com/solutions/Microbiology/tryptose-number-2