Learn More
MOLTOX Molecular Toxicology Inc Tryptose Phosphate Broth, 100 mL/bottle, Store at Room Temperature
SDP
Catalog No. 50948779
Encompass_Preferred
Tryptone Phosphate Broth is used for the cultivation of fastidous, pathogenic microorganisms. Each lot of media from MOLTOX is tested for sterility, where applicable, and its ability to support the growth of the appropriate microorganism. All media are accompanied by GLP-compliant formulation and Quality Control certificate.
Catalog No. 50-948-779 Supplier MOLTOX Molecular Toxicology Inc Supplier No. 26615
Tryptone Phosphate Broth is used for the cultivation of fastidous, pathogenic microorganisms. Each lot of media from MOLTOX is tested for sterility, where applicable, and its ability to support the growth of the appropriate microorganism. All media are accompanied by GLP-compliant formulation and Quality Control certificate.