Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Research Products International Corp RPI Tryptose Phosphate Broth, 500 Grams
SDP

Catalog No. 501977946
Encompass_Preferred

Dissolve 29.5 grams in 1 liter of distilled water. Sterilize by autoclaving for 15 minutes.

Formulation:

Digest Peptone: 20.0 g/L
Disodium Phosphate: 2.5 g/L
Dextrose: 2.0 g/L
Sodium Chloride: 5.0 g/L

Catalog No. 50-197-7946 Supplier Research Products International Corp Supplier No. T65200500.0
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Dissolve 29.5 grams in 1 liter of distilled water. Sterilize by autoclaving for 15 minutes.

Formulation:

Digest Peptone: 20.0 g/L
Disodium Phosphate: 2.5 g/L
Dextrose: 2.0 g/L
Sodium Chloride: 5.0 g/L

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.