Learn More
NEOGEN Tryptose Phosphate Broth, 500 g
SDP
Catalog No. 502015579
Encompass_Preferred
Tryptose Phosphate Broth is used for the cultivation of a wide variety of fastidious microorganisms and is not intended for use in the diagnosis of disease or other conditions in humans. This is a versatile, nutritionally rich buffered glucose broth. The medium is a general-purpose broth that has been used as a blood culture medium and with the addition of sodium azide 0.025% as a selective medium for streptococci in dairy products.For more information visit https://www.neogen.com/solutions/Microbiology/tryptose-phosphate-broth
Catalog No. 50-201-5579 Supplier NEOGEN Supplier No. 700003396
Tryptose Phosphate Broth is used for the cultivation of a wide variety of fastidious microorganisms and is not intended for use in the diagnosis of disease or other conditions in humans. This is a versatile, nutritionally rich buffered glucose broth. The medium is a general-purpose broth that has been used as a blood culture medium and with the addition of sodium azide 0.025% as a selective medium for streptococci in dairy products.For more information visit https://www.neogen.com/solutions/Microbiology/tryptose-phosphate-broth