Learn More
Teknova Tryptose Phosphate Broth. 500ml, Sterile. 4 pack.
SDP
Catalog No. 502387789
Encompass
2.00% Tryptose, 0.20% Glucose, 0.50% Sodium Chloride, 0.25% Sodium Phosphate, Dibasic
Catalog No. 50-238-7789 Supplier Teknova Supplier No. T08004PK
2.00% Tryptose, 0.20% Glucose, 0.50% Sodium Chloride, 0.25% Sodium Phosphate, Dibasic