Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ Tryptose Phosphate Broth
GREENER_CHOICE

Catalog No. 18050039
Encompass_Preferred
Change view
Click to view available options
Quantity:
100 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
18-050-039 100 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 18-050-039 Supplier Invitrogen™ Supplier No. 18050039
Add to Cart
Edge
Add to Cart

missing translation for 'validMainframeMsg'

Cell culture supplement for BHK-21 cells used for vaccine production

  • Tryptose Phosphate Broth contains tryptose (an enzymatic hydrolysis of protein), dextrose, sodium chloride and disodium phosphate
  • Manufactured at a cGMP compliant facility, located in Paisley, Scotland, UK, registered with the FDA as a medical device manufacturer and certified to the ISO 13485 standard

Order Info

Shipping Condition: Room Temperature

TRUSTED_SUSTAINABILITY

Specifications

Content And Storage Storage conditions: 2 – 8°C
Shipping conditions: Ambient
Shelf life: 12 months from date of manufacture
Cell Type Mammalian Cells
Product Type Tryptose Phosphate Broth
Quantity 100 mL
Shipping Condition Room Temperature
What is the concentration of tryptose phosphate solution in Tryptose Phosphate Broth (Cat. No. 18050039)?

The Tryptose Phosphate Broth contains 29.5 g/L tryptose phosphate.

For Research Use Only. Not for use in diagnostic procedures.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.