Learn More
HIMEDIA Tryptose, RM030, Microbiology Culture Media Ingredients, Peptone, Enzymatic Protein Hydrolysate, Fastidious Organisms, Vaccine Media, Powder, 500 g
Catalog No. NC4013786
Encompass_Preferred
Tryptose is a specially prepared enzymatic protein hydrolysate formulated to meet the nutritional needs of fastidious micro-organisms. It supplies a rich mix of peptides and amino acids that keeps blood cells in excellent condition, so haemolytic reactions can be clearly demonstrated in tryptose-based media. It is also valued in vaccine and toxin production media for rapid, luxuriant growth and as a supplement that enriches tissue culture media. Dehydrated powder, store in a cool dry place. Packaged as 500 g.
Catalog No. NC4013786 Supplier HIMEDIA Supplier No. RM030500G
Tryptose is a specially prepared enzymatic protein hydrolysate formulated to meet the nutritional needs of fastidious micro-organisms. It supplies a rich mix of peptides and amino acids that keeps blood cells in excellent condition, so haemolytic reactions can be clearly demonstrated in tryptose-based media. It is also valued in vaccine and toxin production media for rapid, luxuriant growth and as a supplement that enriches tissue culture media. Dehydrated powder, store in a cool dry place. Packaged as 500 g.