Learn More
MilliporeSigma Soybean Casein Digest Broth (TSB) in bottle with screw cap and septum 100ML 12/pk
Catalog No. NC9369809
Encompass
Soybean Case in Digest Broth (TSB); Screw cap with septum; 100mL; pH at 25deg.C: 7.3 +/- 0.2; Color: Tan to gold, clear; Incubation Temp: 20deg. to 25deg.C; Application: Sterility testing; Storage: 12deg. to 25deg.C
Catalog No. NC9369809 Supplier MilliporeSigma Supplier No. STBMTSB12
Soybean Case in Digest Broth (TSB); Screw cap with septum; 100mL; pH at 25deg.C: 7.3 +/- 0.2; Color: Tan to gold, clear; Incubation Temp: 20deg. to 25deg.C; Application: Sterility testing; Storage: 12deg. to 25deg.C