Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

MilliporeSigma™ CT-2A Mouse Glioma Cell Line

Catalog No. SCC194 Shop All MilliporeSigma Products
Change view
Click to view available options
Quantity:
≥ 1 x 106 cells/vial
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
SCC194 ≥ 1 x 106 cells/vial
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. SCC194 Supplier MilliporeSigma™ Supplier No. SCC194
Add to Cart
Add to Cart

CT-2A mouse glioma cell line is a well characterized and highly published model for breast cancer research.

  • Metastasizing (stage IV) breast cancers have the lowest survival rate among all stages of breast cancer
  • Metastases may take on a range of morphologies in their new locations, complicating diagnosis and decisions on courses of treatment
  • Tumor cell lines that mimic the in vivo behavior of metastasizing cancers are thus critical tools in breast cancer research
  • The cells can be cultured for at least 10 passages after initial thawing without significantly affecting the cell marker expression and functionality
  • Cells are tested negative for infectious diseases by a Mouse Essential CLEAR panel by Charles River Animal Diagnostic Services
  • Cells are verified to be of mouse origin and negative for inter-species contamination from rat, Chinese hamster, Golden Syrian hamster, human and non-human primate (NHP) as assessed by a Contamination CLEAR panel by Charles River Animal Diagnostic Services
  • Cells are negative for mycoplasma contamination
TRUSTED_SUSTAINABILITY

Specifications

Content And Storage Store in liquid nitrogen
Format Vial
Quantity ≥ 1 x 106 cells/vial
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.