Learn More
Foxx Life Sciences 250mL PETG Media Bottle Non Sterile, No Cap, 60CS
Catalog No. 502551682
Encompass_Preferred
PETG Bottles are supplied as endotoxin free, nuclease free, USP 665 and USP 788. The Non Sterile Media Bottles with 38-430 neck are double bagged and manufactured in a cleanroom for bioprocessing applications. 250mL PETG bottles are leakproof, clear, designed for packaging equipment, freezer safe, and made of USP Class VI Resins. These laboratory PETG Bottles are designed for buffers, storing reagents, PH sensitive materials, and biopharmaceutical solutions. Our new low cost EZBio Titanium Square Bottles are sourced from our Foxx Life Sciences India Plant.
Catalog No. 50-255-1682 Supplier Foxx Life Sciences Supplier No. 6273003FLS
PETG Bottles are supplied as endotoxin free, nuclease free, USP 665 and USP 788. The Non Sterile Media Bottles with 38-430 neck are double bagged and manufactured in a cleanroom for bioprocessing applications. 250mL PETG bottles are leakproof, clear, designed for packaging equipment, freezer safe, and made of USP Class VI Resins. These laboratory PETG Bottles are designed for buffers, storing reagents, PH sensitive materials, and biopharmaceutical solutions. Our new low cost EZBio Titanium Square Bottles are sourced from our Foxx Life Sciences India Plant.