Learn More
Foxx Life Sciences 500mL PETG Media Bottle Non Sterile, No Cap, 48CS
GREENER_CHOICE
Catalog No. 502551680
Encompass_Preferred
PETG Bottles are supplied as endotoxin free, nuclease free, USP 665 and USP 788. The Non Sterile Media Bottles with 38-430 neck are double bagged and manufactured in a cleanroom for bioprocessing applications. 500mL PETG bottles are leakproof, clear, designed for packaging equipment, freezer safe, and made of USP Class VI Resins. These laboratory PETG Bottles are designed for buffers, storing reagents, PH sensitive materials, and biopharmaceutical solutions. Our new low cost EZBio Titanium Square Bottles are sourced from our Foxx Life Sciences India Plant.
Catalog No. 50-255-1680 Supplier Foxx Life Sciences Supplier No. 6274003FLS
PETG Bottles are supplied as endotoxin free, nuclease free, USP 665 and USP 788. The Non Sterile Media Bottles with 38-430 neck are double bagged and manufactured in a cleanroom for bioprocessing applications. 500mL PETG bottles are leakproof, clear, designed for packaging equipment, freezer safe, and made of USP Class VI Resins. These laboratory PETG Bottles are designed for buffers, storing reagents, PH sensitive materials, and biopharmaceutical solutions. Our new low cost EZBio Titanium Square Bottles are sourced from our Foxx Life Sciences India Plant.