Learn More
Foxx Life Sciences 125mL PETG Media Bottle Sterile, 96CS
GREENER_CHOICE
Catalog No. 502551683
Encompass_Preferred
PETG Bottles are supplied as endotoxin free, nuclease free, USP 665, USP 788, and Gamma Sterilized 10-6. The Sterile Media Bottles with 38-430 neck are double bagged and manufactured in a cleanroom for bioprocessing applications. 125mL PETG bottles are leakproof, clear, designed for packaging equipment, freezer safe, and made of USP Class VI Resins. These laboratory PETG Bottles are designed for buffers, storing reagents, PH sensitive materials, and biopharmaceutical solutions. Our new low cost EZBio Titanium Square Bottles are sourced from our Foxx Life Sciences India Plant.
Catalog No. 50-255-1683 Supplier Foxx Life Sciences Supplier No. 6272101FLS
PETG Bottles are supplied as endotoxin free, nuclease free, USP 665, USP 788, and Gamma Sterilized 10-6. The Sterile Media Bottles with 38-430 neck are double bagged and manufactured in a cleanroom for bioprocessing applications. 125mL PETG bottles are leakproof, clear, designed for packaging equipment, freezer safe, and made of USP Class VI Resins. These laboratory PETG Bottles are designed for buffers, storing reagents, PH sensitive materials, and biopharmaceutical solutions. Our new low cost EZBio Titanium Square Bottles are sourced from our Foxx Life Sciences India Plant.