Learn More
Selleck Chemical LLC TSA (Trichostatin A) S1045-5mg
SDP
Catalog No. 503108138
Encompass_Preferred
TSA (Trichostatin A) is an HDAC inhibitor with IC50 of 1 8 nM in cell-free assays
Catalog No. 50-310-8138 Supplier Selleck Chemical LLC Supplier No. S10455MG
TSA (Trichostatin A) is an HDAC inhibitor with IC50 of 1 8 nM in cell-free assays