Learn More
General Laboratory Products Tryptic Soy Agar (TSA), General-Purpose Culture Medium, 200 ml
Catalog No. NC2684467
Encompass_Preferred
Tryptic Soy Agar (TSA) is a versatile, general-purpose growth medium in a 200 ml easy-pour narrow-mouth glass bottle, perfect for isolating and cultivating a wide range of microorganisms, including bacteria, yeast, and fungi. Packaged as 9 per case, it supports both fastidious and non-fastidious organisms, ensuring robust microbial growth. With a pH of 7.3 ± 0.2, it offers consistent performance, making it an essential tool for microbiological applications.
Catalog No. NC2684467 Supplier General Laboratory Products Supplier No. 250EZ3700200
Tryptic Soy Agar (TSA) is a versatile, general-purpose growth medium in a 200 ml easy-pour narrow-mouth glass bottle, perfect for isolating and cultivating a wide range of microorganisms, including bacteria, yeast, and fungi. Packaged as 9 per case, it supports both fastidious and non-fastidious organisms, ensuring robust microbial growth. With a pH of 7.3 ± 0.2, it offers consistent performance, making it an essential tool for microbiological applications.