Learn More
Teknova TSA Plates Lec-Tween80 10pk
SDP
Catalog No. 5001082695
Encompass
Tryptic Soy Agar Settle Plates with Lecithin and Tween-80 100mm 10 Plates/Sleeve Sterile
Catalog No. 50-010-82695 Supplier Teknova Supplier No. 23914
Tryptic Soy Agar Settle Plates with Lecithin and Tween-80 100mm 10 Plates/Sleeve Sterile