Learn More
Teknova TSA Plates Lec-Tween80 5pk
SDP
Catalog No. 5001082696
Encompass
Tryptic Soy Agar Settle Plates with Lecithin and Tween-80 100mm 5 Plates/Sleeve Sterile
Catalog No. 50-010-82696 Supplier Teknova Supplier No. 23915
Tryptic Soy Agar Settle Plates with Lecithin and Tween-80 100mm 5 Plates/Sleeve Sterile