Learn More
Quanterix Inc TSA PLUS CYANINE 3 SYSTEM
SDP
Catalog No. 509049411
Encompass
TSA Plus Cyanine 3 System, for 50-150 Slides. These systems are designed for direct fluorescence detection in ISH or IHC procedures.
Catalog No. 50-904-9411 Supplier Quanterix Inc Supplier No. NEL744001KT
TSA Plus Cyanine 3 System, for 50-150 Slides. These systems are designed for direct fluorescence detection in ISH or IHC procedures.