Learn More
Quanterix Inc TSA PLUS CYANINE 5 SYSTEM
SDP
Catalog No. 509049413
Encompass
TSA Plus Cyanine 5 System, for 50-150 Slides. These systems are designed for direct fluorescence detection in ISH or IHC procedures.
Catalog No. 50-904-9413 Supplier Quanterix Inc Supplier No. NEL745001KT
TSA Plus Cyanine 5 System, for 50-150 Slides. These systems are designed for direct fluorescence detection in ISH or IHC procedures.